Assay Details
| Gene Symbol: MTFR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716109.3 | XP_002716155.1 | 6 - 7 | 987 | 89 | |
| XM_008265923.2 | XP_008264145.1 | 6 - 7 | 952 | 89 | ||
| XM_017346584.1 | XP_017202073.1 | 6 - 7 | 977 | 89 | ||
More Information
Set Membership: |
|

MTFR1L
Genomic Map