Assay Details
| Gene Symbol: CHMP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721899.3 | XP_002721945.1 | 730 | 62 | ||
| XM_008249035.2 | XP_008247257.1 | 667 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHMP2A| Gene Symbol: CHMP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721899.3 | XP_002721945.1 | 730 | 62 | ||
| XM_008249035.2 | XP_008247257.1 | 667 | 62 | |||
Set Membership: |
|