Assay Details
| Gene Symbol: TMEM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715971.3 | XP_002716017.1 | 4 - 5 | 546 | 64 | |
| XM_008265656.2 | XP_008263878.1 | 3 - 4 | 417 | 64 | ||
More Information
Set Membership: |
|

TMEM50A
Genomic Map