Assay Details
| Gene Symbol: MYOZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717224.3 | XP_002717270.1 | 4 - 5 | 554 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYOZ2| Gene Symbol: MYOZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717224.3 | XP_002717270.1 | 4 - 5 | 554 | 81 | |
Set Membership: |
|