Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712763.3 | XP_002712809.1 | 10 - 11 | 1416 | 63 | |
| XM_008259658.2 | XP_008257880.1 | 10 - 11 | 1387 | 63 | ||
More Information
Set Membership: |
|

WASHC1
Genomic Map