Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259658.2 | XP_008257880.1 | 1 - 2 | 101 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008259658.2 transcripts.
Set Membership: |
|

WASHC1
Genomic Map