Assay Details
| Gene Symbol: KNSTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269196.2 | XP_008267418.1 | 6 - 7 | 757 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KNSTRN| Gene Symbol: KNSTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269196.2 | XP_008267418.1 | 6 - 7 | 757 | 57 | |
Set Membership: |
|