Assay Details
| Gene Symbol: POLD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708695.3 | XP_002708741.1 | 4 - 5 | 378 | 63 | |
| XM_008264043.2 | XP_008262265.1 | 4 - 5 | 378 | 63 | ||
More Information
Set Membership: |
|

POLD3
Genomic Map