Assay Details
| Gene Symbol: LSMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260685.2 | XP_008258907.1 | 1 - 2 | 936 | 107 | |
| XM_008260687.2 | XP_008258909.1 | 1 - 2 | 936 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008260685.2, XM_008260687.2 transcripts.
Set Membership: |
|

LSMEM2
Genomic Map