Assay Details
| Gene Symbol: LSMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260685.2 | XP_008258907.1 | 2 - 3 | 1036 | 78 | |
| XM_008260687.2 | XP_008258909.1 | 2 - 3 | 1036 | 78 | ||
More Information
Set Membership: |
|

LSMEM2
Genomic Map