Assay Details
| Gene Symbol: ZKSCAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721860.3 | XP_002721906.1 | 1071 | 65 | ||
| XM_008248902.2 | XP_008247124.1 | 894 | 65 | |||
| XM_008248903.2 | XP_008247125.1 | 1146 | 65 | |||
| XM_017337858.1 | XP_017193347.1 | 1323 | 65 | |||
More Information
Set Membership: |
|

ZKSCAN1
Genomic Map