Assay Details
| Gene Symbol: IGF2BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261527.2 | XP_008259749.1 | 14 | 3480 | 121 | |
| XM_017344378.1 | XP_017199867.1 | 14 | 3477 | 121 | ||
More Information
Set Membership: |
|

IGF2BP3
Genomic Map