Assay Details
| Gene Symbol: LOC103344959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255934.2 | XP_008254156.1 | 205 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008255934.2 transcripts.
Set Membership: |
|

LOC103344959
Genomic Map