Assay Details
| Gene Symbol: FAM111A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271647.2 | XP_008269869.1 | 2 - 3 | 429 | 103 | |
| XM_008271656.2 | XP_008269878.2 | 2 - 3 | 509 | 103 | ||
| XM_017346696.1 | XP_017202185.1 | 3 - 4 | 367 | 103 | ||
More Information
Set Membership: |
|

FAM111A
Genomic Map