Assay Details
| Gene Symbol: B3GNTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251182.2 | XP_008249404.1 | 755 | 55 | ||
| XM_008251183.2 | XP_008249405.1 | 755 | 55 | |||
| XM_008251184.2 | XP_008249406.1 | 755 | 55 | |||
| XM_008251185.2 | XP_008249407.1 | 755 | 55 | |||
| XM_017339052.1 | XP_017194541.1 | 755 | 55 | |||
| XM_017339053.1 | XP_017194542.1 | 755 | 55 | |||
| XM_017339055.1 | XP_017194544.1 | 745 | 55 | |||
More Information
Set Membership: |
|

B3GNTL1
Genomic Map