Assay Details
| Gene Symbol: IGFLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249531.2 | XP_008247753.1 | 1718 | 53 | ||
| XM_008249532.2 | XP_008247754.1 | 916 | 53 | |||
| XM_017338170.1 | XP_017193659.1 | 641 | 53 | |||
More Information
Set Membership: |
|

IGFLR1
Genomic Map