Assay Details
| Gene Symbol: IRAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249927.2 | XP_008248149.1 | 703 | 65 | ||
| XM_008249928.2 | XP_008248150.1 | 701 | 65 | |||
| XM_008249929.2 | XP_008248151.1 | 703 | 65 | |||
| XM_017338407.1 | XP_017193896.1 | 521 | 65 | |||
| XM_017338408.1 | XP_017193897.1 | 707 | 65 | |||
More Information
Set Membership: |
|

IRAK2
Genomic Map