Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714489.3 | XP_002714535.1 | 2 - 3 | 605 | 79 | |
| XM_008263026.2 | XP_008261248.1 | 2 - 3 | 545 | 79 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map