Assay Details
| Gene Symbol: LOC100345427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714462.3 | XP_002714508.2 | 10 - 11 | 1478 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100345427| Gene Symbol: LOC100345427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714462.3 | XP_002714508.2 | 10 - 11 | 1478 | 100 | |
Set Membership: |
|