Assay Details
| Gene Symbol: LOC100345427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714462.3 | XP_002714508.2 | 1 - 2 | 414 | 87 | |
| XM_017345162.1 | XP_017200651.1 | 1 - 2 | 416 | 87 | ||
More Information
Set Membership: |
|

LOC100345427
Genomic Map