Assay Details
| Gene Symbol: LOC100345427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714462.3 | XP_002714508.2 | 9 - 10 | 1390 | 71 | |
| XM_017345162.1 | XP_017200651.1 | 9 - 10 | 1392 | 71 | ||
More Information
Set Membership: |
|

LOC100345427
Genomic Map