Assay Details
| Gene Symbol: TP53I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710069.3 | XP_002710115.1 | 3 - 4 | 528 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TP53I3| Gene Symbol: TP53I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710069.3 | XP_002710115.1 | 3 - 4 | 528 | 64 | |
Set Membership: |
|