Assay Details
| Gene Symbol: HAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715457.3 | XP_002715503.3 | 6 - 7 | 851 | 54 | |
| XM_008264383.2 | XP_008262605.1 | 6 - 7 | 859 | 54 | ||
More Information
Set Membership: |
|

HAX1
Genomic Map