Assay Details
| Gene Symbol: SIGMAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708016.3 | XP_002708062.1 | 2 - 3 | 469 | 90 | |
| XM_008272727.2 | XP_008270949.1 | 2 - 3 | 475 | 90 | ||
More Information
Set Membership: |
|

SIGMAR1
Genomic Map