Assay Details
| Gene Symbol: MST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260652.2 | XP_008258874.1 | 6 - 7 | 744 | 51 | |
| XM_008260653.2 | XP_008258875.2 | 6 - 7 | 645 | 51 | ||
More Information
Set Membership: |
|

MST1
Genomic Map