Assay Details
Gene Symbol: C3H5orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008255027.2 | XP_008253249.1 | 1 - 2 | 235 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008255027.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
