Assay Details
| Gene Symbol: LOC100343920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717544.3 | XP_002717590.1 | 10 - 11 | 1699 | 85 | |
| XM_008268483.2 | XP_008266705.1 | 10 - 11 | 1536 | 85 | ||
| XM_008268485.2 | XP_008266707.1 | 10 - 11 | 1527 | 85 | ||
| XM_008268489.2 | XP_008266711.1 | 9 - 10 | 1347 | 85 | ||
| GenBank mRNA | Z35490.1 | - | 1639 | 85 | ||
More Information
Set Membership: |
|

LOC100343920
Genomic Map