Assay Details
| Gene Symbol: LOC100343920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717544.3 | XP_002717590.1 | 6 - 7 | 952 | 85 | |
| XM_008268483.2 | XP_008266705.1 | 6 - 7 | 789 | 85 | ||
| XM_008268485.2 | XP_008266707.1 | 6 - 7 | 780 | 85 | ||
| XM_008268489.2 | XP_008266711.1 | 6 - 7 | 789 | 85 | ||
| XM_008268490.2 | XP_008266712.1 | 6 - 7 | 789 | 85 | ||
| XM_017347823.1 | XP_017203312.1 | 6 - 7 | 789 | 85 | ||
| GenBank mRNA | Z35490.1 | - | 892 | 85 | ||
More Information
Set Membership: |
|

LOC100343920
Genomic Map