Assay Details
| Gene Symbol: LOC100343920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268489.2 | XP_008266711.1 | 8 - 9 | 1165 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100343920| Gene Symbol: LOC100343920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268489.2 | XP_008266711.1 | 8 - 9 | 1165 | 100 | |
Set Membership: |
|