Assay Details
| Gene Symbol: ST6GAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716497.3 | XP_002716543.1 | 7 - 8 | 1646 | 57 | |
| XM_017346979.1 | XP_017202468.1 | 8 - 9 | 1757 | 57 | ||
More Information
Set Membership: |
|

ST6GAL1
Genomic Map