Assay Details
| Gene Symbol: BHLHB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349650.1 | XP_017205139.1 | 8 | 2110 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BHLHB9| Gene Symbol: BHLHB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349650.1 | XP_017205139.1 | 8 | 2110 | 95 | |
Set Membership: |
|