Assay Details
| Gene Symbol: CERT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251138.2 | XP_008249360.1 | 1291 | 72 | ||
| XM_017339030.1 | XP_017194519.1 | 1291 | 72 | |||
| XM_017339031.1 | XP_017194520.1 | 1291 | 72 | |||
More Information
Set Membership: |
|

CERT1
Genomic Map