Assay Details
| Gene Symbol: C7H2orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343193.1 | XP_017198682.1 | 2 - 3 | 485 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7H2orf72| Gene Symbol: C7H2orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343193.1 | XP_017198682.1 | 2 - 3 | 485 | 100 | |
Set Membership: |
|