Assay Details
| Gene Symbol: LOC100354911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347437.1 | XP_017202926.1 | 5 - 6 | 427 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100354911| Gene Symbol: LOC100354911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347437.1 | XP_017202926.1 | 5 - 6 | 427 | 99 | |
Set Membership: |
|