Assay Details
| Gene Symbol: NAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256816.2 | XP_008255038.2 | 11 - 12 | 1013 | 68 | |
| XM_008256819.2 | XP_008255041.1 | 11 - 12 | 1010 | 68 | ||
| XM_008256820.2 | XP_008255042.1 | 11 - 12 | 1013 | 68 | ||
| XM_008256821.2 | XP_008255043.1 | 9 - 10 | 821 | 68 | ||
| XM_017341988.1 | XP_017197477.1 | 11 - 12 | 1013 | 68 | ||
More Information
Set Membership: |
|

NAP1L1
Genomic Map