Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719346.3 | XP_002719392.1 | 2 - 3 | 346 | 61 | |
| XM_008271421.2 | XP_008269643.1 | 2 - 3 | 350 | 61 | ||
| XM_008271424.2 | XP_008269646.1 | 2 - 3 | 361 | 61 | ||
More Information
Set Membership: |
|

ORMDL3
Genomic Map