Assay Details
| Gene Symbol: HINT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002707996.3 | XP_002708042.1 | 4 - 5 | 420 | 60 | |
| XM_008268271.2 | XP_008266493.1 | 3 - 4 | 1389 | 60 | ||
| XM_008268313.2 | XP_008266535.1 | 4 - 5 | 1145 | 60 | ||
| XM_008268369.2 | XP_008266591.1 | 4 - 5 | 587 | 60 | ||
| XM_008268465.2 | XP_008266687.1 | 2 - 3 | 1296 | 60 | ||
| XM_008268518.2 | XP_008266740.1 | 4 - 5 | 961 | 60 | ||
More Information
Set Membership: |
|

HINT2
Genomic Map