Assay Details
| Gene Symbol: TECRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717154.3 | XP_002717200.1 | 10 - 11 | 981 | 71 | |
| XM_008267813.2 | XP_008266035.1 | 10 - 11 | 982 | 71 | ||
| XM_017347532.1 | XP_017203021.1 | 8 - 9 | 865 | 71 | ||
| XM_017347533.1 | XP_017203022.1 | 8 - 9 | 793 | 71 | ||
More Information
Set Membership: |
|

TECRL
Genomic Map