Assay Details
| Gene Symbol: MLIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263062.2 | XP_008261284.1 | 12 - 13 | 2989 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLIP| Gene Symbol: MLIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263062.2 | XP_008261284.1 | 12 - 13 | 2989 | 64 | |
Set Membership: |
|