Assay Details
| Gene Symbol: PPP1R3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718505.3 | XP_002718551.3 | 1 - 2 | 133 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002718505.3 transcripts.
Set Membership: |
|

PPP1R3C
Genomic Map