Assay Details
| Gene Symbol: PMP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718844.3 | XP_002718890.1 | 4 - 5 | 563 | 53 | |
| XM_008270703.2 | XP_008268925.1 | 4 - 5 | 538 | 53 | ||
More Information
Set Membership: |
|

PMP22
Genomic Map