Assay Details
| Gene Symbol: LOC100344472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711877.3 | XP_002711923.1 | 18 - 19 | 2555 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100344472| Gene Symbol: LOC100344472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711877.3 | XP_002711923.1 | 18 - 19 | 2555 | 72 | |
Set Membership: |
|