Assay Details
| Gene Symbol: CILK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263058.2 | XP_008261280.1 | 8 - 9 | 1249 | 63 | |
| XM_017345200.1 | XP_017200689.1 | 9 - 10 | 1452 | 63 | ||
More Information
Set Membership: |
|

CILK1
Genomic Map