Assay Details
| Gene Symbol: TRAPPC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721126.3 | XP_002721172.1 | 1742 | 61 | ||
| XM_008274553.2 | XP_008272775.1 | 1732 | 61 | |||
| XM_008274555.2 | XP_008272777.1 | 1362 | 61 | |||
More Information
Set Membership: |
|

TRAPPC12
Genomic Map