Assay Details
Gene Symbol: NRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259660.2 | XP_008257882.1 | 5 - 6 | 1143 | 56 | |
XM_008259661.2 | XP_008257883.1 | 5 - 6 | 623 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
