Assay Details
| Gene Symbol: LGALS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257257.2 | XP_008255479.1 | 3 - 4 | 394 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LGALS7| Gene Symbol: LGALS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257257.2 | XP_008255479.1 | 3 - 4 | 394 | 75 | |
Set Membership: |
|