Assay Details
| Gene Symbol: POP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257990.2 | XP_008256212.1 | 1 - 2 | 114 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008257990.2 transcripts.
Set Membership: |
|

POP7
Genomic Map