Assay Details
| Gene Symbol: LOC100353962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719416.3 | XP_002719462.1 | 1 - 2 | 151 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002719416.3 transcripts.
Set Membership: |
|

LOC100353962
Genomic Map