Assay Details
| Gene Symbol: LOC100353962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719416.3 | XP_002719462.1 | 5 - 6 | 658 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100353962| Gene Symbol: LOC100353962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719416.3 | XP_002719462.1 | 5 - 6 | 658 | 103 | |
Set Membership: |
|