Assay Details
| Gene Symbol: MTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718766.3 | XP_002718812.1 | 1 - 2 | 179 | 54 | |
| XM_008270640.2 | XP_008268862.1 | 1 - 2 | 180 | 54 | ||
| XM_008270642.2 | XP_008268864.1 | 1 - 2 | 180 | 54 | ||
| XM_008270643.2 | XP_008268865.1 | 1 - 2 | 180 | 54 | ||
More Information
Set Membership: |
|

MTG1
Genomic Map